Kalciumsulfat, Drierite™ 10-20 mesh för torkning, with indicator (cobalt (II) chloride)
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Calcium sulphate (anhydrous) anhydrite, Gypsum
43063.TD 43063.XA
43063.TDEA 1625 SEK
43063.TD 43063.XA
Kalciumsulfat, Drierite™ 10-20 mesh för torkning, with indicator (cobalt (II) chloride)
Kalciumsulfat
CAS No.: 7778-18-9 Physical Form: Granular, -10+20 Mesh Sensitivity: Hygroscopic Appearance: Blue to pink Odour: Odourless Merck Reference: 14,1706 MDL No.: MFCD00010912
Formel: CaO₄S
MW: 136.14 g/mol
Smältpunkt: 1350
Storage Temperature: Omgivande
MDL Number: MFCD00010912
CAS nummer: 7778-18-9
EINECS: 231-900-3
UN: 3077
ADR: 9 III
Merck Index: 14,1706