2-Chloro-5-pyridineboronic acid (contains varying amounts of Anhydride)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Chloropyridine-5-boronic acid, 2-Chloro-5-pyridinylboronic acid, 6-Chloro-3-pyridinylboronic acid, (6-Chloropyridin-3-yl)boronic acid, 6-Chloropyridine-3-boronic acid
C2288-1G
C2288-5G
TCIAC2288-1GEA
605
SEK
TCIAC2288-1G
TCIAC2288-5G
2-Chloro-5-pyridineboronic acid (contains varying amounts of Anhydride)
2-Chloro-5-pyridineboronic acid
Formel:
C₅H₅BClNO₂ MW: 157,36 g/mol Smältpunkt: 167°C |
MDL Number:
MFCD03094998 CAS nummer: 444120-91-6 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (Neutralization titration) | 96.0 to 113.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...