p-Tolylacetonitrile ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: P-Toluylacetonitrile, 4-Methylbenzyl cyanide, (4-Methylphenyl)acetonitrile

X0047-25ML X0047-500ML
TCIAX0047-25MLEA 350 SEK
TCIAX0047-25ML TCIAX0047-500ML
p-Tolylacetonitrile ≥98.0%
p-Tolylacetonitrile
Formel: C₉H₉N
MW: 131,18 g/mol
Kokpunkt: 245°C
Smältpunkt: 18 °C
Densitet: 0,99 g/cm³ (20 °C)
Flampunkt: 106°C
MDL Number: MFCD00001922
CAS nummer: 2947-61-7
EINECS: 220-963-2
UN: 3276
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b53d1a6eb92291df0cb19b8978ec7042ce2ca4d&vid=9b53d1a6eb92291df0cb19b8978ec7042ce2ca4d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR