Etylmetakrylat 99,0 GC_ANALYSMETOD Stabilised

Leverantör: TCI
Fara

Synonyms: Ethyl 2-methylpropenoate, Methacrylic acid ethyl ester, Ethyl-2-methylprop-2-enoat, Ethyl-2-methylpropenoat, Ethyl 2-methylprop-2-enoate

Stabilization: Hydrochinon

M0084-25ML M0084-500ML
TCIAM0084-25MLEA 215 SEK
TCIAM0084-25ML TCIAM0084-500ML
Etylmetakrylat 99,0 GC_ANALYSMETOD Stabilised
Etylmetakrylat
Formel: C₆H₁₀O₂
MW: 114.14 g/mol
Kokpunkt: 117°C
Smältpunkt: –75°C
Flampunkt: 20°C
MDL Number: MFCD00009161
CAS nummer: 97-63-2
EINECS: 202-597-5
UN: 2277
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d87e0e9b934ff6d0e6c212e976ea4e5303d731d&vid=5d87e0e9b934ff6d0e6c212e976ea4e5303d731d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR