Thiophene-2-carbaldehyde ≥98.0% stabilised
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Thiophenecarboxaldehyde, Thiophene-2-carboxaldehyde
Stabilization: med hydrokinon
T0725-25ML T0725-500ML T0725-100ML
TCIAT0725-25MLEA 535 SEK
TCIAT0725-25ML TCIAT0725-500ML TCIAT0725-100ML
Thiophene-2-carbaldehyde ≥98.0% stabilised
Thiophene-2-carbaldehyde
Formel: C₅H₄OS
MW: 112,15 g/mol
Kokpunkt: 198°C
Smältpunkt: ? 10 °C
Densitet: 1,2264 g/cm³ (20 °C)
Flampunkt: 78°C
Storage Temperature: Kylskåp
MDL Number: MFCD00005429
CAS nummer: 98-03-3
EINECS: 202-629-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=330bc0fa1bf25c05ef226d85dbc43e7f0afd9862&vid=330bc0fa1bf25c05ef226d85dbc43e7f0afd9862&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %