2-Phenoxyaniline ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
Synonyms:
1-Amino-2-phenoxybenzol
A1142-25G
A1142-500G
TCIAA1142-25GEA
405
SEK
TCIAA1142-25G
TCIAA1142-500G
2-Phenoxyaniline ≥98.0% (by GC, titration analysis)
2-Phenoxyaniline
Formel:
C₁₂H₁₁NO Kokpunkt: 187°C Smältpunkt: 46…48 °C Storage Temperature: Kylskåp |
MDL Number:
MFCD00035765 CAS nummer: 2688-84-8 EINECS: 220-254-8 |
|
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...