(±)-Ethyl-2-methylbutyrate 98,0 GC_ANALYSMETOD
Leverantör: TCI
Synonyms:
(±)-2-Methylbutanoic acid ethyl ester, (±)-2-Methylbutyric acid ethyl ester, Ethyl 2-methylbutanoate
M0757-25ML
M0757-500ML
TCIAM0757-25MLEA
215
SEK
TCIAM0757-25ML
TCIAM0757-500ML
(±)-Ethyl-2-methylbutyrate 98,0 GC_ANALYSMETOD
(±)-Ethyl-2-methylbutyrate
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...