Copper(II) acetylacetonate ≥97.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Bis(2,4-pentanedionato-O,O')copper (II), Copper (II) bis(2,4-pentanedionato-O,O'), Bis(acetylacetone)copper, Copper (II) 2,4-pentanedionate

C0384-25G C0384-250G
TCIAC0384-25GEA 230 SEK
TCIAC0384-25G TCIAC0384-250G
Copper(II) acetylacetonate ≥97.0%
Copper(II) acetylacetonate
Formel: C₁₀H₁₄CuO₄
MW: 261,76 g/mol
Smältpunkt: 279…283 °C
MDL Number: MFCD00000016
CAS nummer: 13395-16-9
EINECS: 236-477-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca83eb08990e0adc8c5116a446a83c1fd588f282&vid=ca83eb08990e0adc8c5116a446a83c1fd588f282&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Blue to Dark blue to Dark purple powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Drying loss max. 0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR