Tetraethylammonium perchlorate ≥98% wetted with 10% water
Leverantör: Thermo Scientific
Fara
Stabilization: med vatten
44062.09 44062.18
44062.09EA 1700 SEK
44062.09 44062.18
Tetraethylammonium perchlorate ≥98% wetted with 10% water
Tetraethylammonium perchlorate
Tetraethylammonium perchlorate is used as supporting electrolyte in polarographic measurements.
Purity: 98% CAS No.: 2567-83-1 Molecular Formula: (C2H5)4NClO4 Formula Weight: 229.71 Physical Form: Wet crystals Odour: Odourless Beilstein Reference: 3638093 UN No.: 1479 MDL No.: MFCD00041977
Formel: C₈H₂₀ClNO₄
MW: 229,7 g/mol
Smältpunkt: >300 °C
Densitet: 1,3 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00041977
CAS nummer: 2567-83-1
EINECS: 219-904-3
UN: 1479
ADR: 5.1,II