2-Nitro-9-fluorenone ≥98.0% (by HPLC, total nitrogen)
Leverantör: TCI EUROPE
Varning
Synonyms: 2-Nitro-9H-fluoren-9-one
N0202-1G N0202-10G
TCIAN0202-1GEA 590 SEK
TCIAN0202-1G TCIAN0202-10G
2-Nitro-9-fluorenone ≥98.0% (by HPLC, total nitrogen)
2-Nitro-9-fluorenone
Formel: C₁₃H₇NO₃
MW: 225,2 g/mol
Kokpunkt: 419 °C (1013 hPa)
Smältpunkt: 224,3 °C
Densitet: 1,44 g/cm³ (20 °C)
MDL Number: MFCD00001152
CAS nummer: 3096-52-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f5b326e3deaac65ebcd08254a47d3cce8c3a90d&vid=7f5b326e3deaac65ebcd08254a47d3cce8c3a90d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Yellow to Orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 220.0 to 224.0 deg-C
Solubility in hot Acetic acid almost transparency