1-(2-Bromoethoxy)-4-nitrobenzene ≥97.0%
Leverantör: TCI EUROPE
Varning
B3254-1G B3254-5G
TCIAB3254-1GEA 1100 SEK
TCIAB3254-1G TCIAB3254-5G
1-(2-Bromoethoxy)-4-nitrobenzene ≥97.0%
1-(2-Bromoethoxy)-4-nitrobenzene
Formel: C₈H₈BrNO₃
MW: 246,06 g/mol
Kokpunkt: 170°C
MDL Number: MFCD00031363
CAS nummer: 13288-06-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d19c394fa1d406d93b1cfe4d9312540330424d8&vid=8d19c394fa1d406d93b1cfe4d9312540330424d8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Melting point 63.0 to 67.0 deg-C