(S)-(-)-3-Butyne-2-ol ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: (S)-3-Butyn-2-ol, (S)-(-)-3-Hydroxy-1-butyne, (S)-(-)-But-3-yn-2-ol
B2910-1G B2910-5G
TCIAB2910-1GEA 870 SEK
TCIAB2910-1G TCIAB2910-5G
(S)-(-)-3-Butyne-2-ol ≥98.0%
(S)-(-)-3-Butyne-2-ol
Formel: C₄H₆O
MW: 70,09 g/mol
Kokpunkt: 100°C
Flampunkt: 26°C
MDL Number: MFCD00190166
CAS nummer: 2914-69-4
UN: 2929
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=071cee0281582e931d11029833b25cfc419b334c&vid=071cee0281582e931d11029833b25cfc419b334c&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%