(R)-(+)-1-Phenylethanol ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
(R)-1-Phenylethanol, (R)-(+)-α-Methylbenzyl alcohol, (R)-(+)-sec-Phenethyl alcohol
P0795-1G
P0795-25G
P0795-5G
TCIAP0795-1GEA
550
SEK
TCIAP0795-1G
TCIAP0795-25G
TCIAP0795-5G
(R)-(+)-1-Phenylethanol ≥98.0%
(R)-(+)-1-Phenylethanol
Formel:
C₈H₁₀O MW: 122.17 g/mol Kokpunkt: 207 Flampunkt: 86°C Storage Temperature: Omgivande |
MDL Number:
MFCD00064263 CAS nummer: 1517-69-7 UN: 2937 ADR: 6.1 III |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...