2-Iodo-1-phenylethane ≥98.0% stabilised

Leverantör: TCI EUROPE

Synonyms: 2-Phenylethyl iodide, (2-Iodoethyl)benzene

Stabilization: Stabilised with copper chip

I0521-5G I0521-1G
TCIAI0521-5GEA 735 SEK
TCIAI0521-5G TCIAI0521-1G
2-Iodo-1-phenylethane ≥98.0% stabilised
2-Iodo-1-phenylethane
Formel: C₆H₅CH₂CH₂I
MW: 232,06 g/mol
Kokpunkt: 123°C
Smältpunkt: –2 °C
Densitet: 1,6323 g/cm³ (20 °C)
MDL Number: MFCD00039408
CAS nummer: 17376-04-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=356e91d8bb8214fdb106caceabfa1f773b05a31f&vid=356e91d8bb8214fdb106caceabfa1f773b05a31f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR