15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
15-Crown-5, 1,4,7,10,13-Pentaoxacyclopentadecane
C0859-25ML
C0859-5ML
TCIAC0859-25MLEA
2950
SEK
TCIAC0859-25ML
TCIAC0859-5ML
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V) ≥97.0%
15-Crown-5 (1,4,7,10,13-pentaoxacyclopentadecane, lead ionophore V)
Formel:
C₁₀H₂₀O₅ MW: 220,27 g/mol Kokpunkt: 116°C Smältpunkt: –20°C Densitet: 1,11 g/cm³ (20 °C) Storage Temperature: Omgivande |
MDL Number:
MFCD00005110 CAS nummer: 33100-27-5 EINECS: 251-379-6 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...