1,4-Cyclohexadiene ≥98.0% stabilised

Leverantör: TCI EUROPE
Fara

Synonyms: Cyclohexa-1,4-diene

Stabilization: Stabilised med BHT/Ionol

C0468-10ML C0468-25ML
TCIAC0468-10MLEA 950 SEK
TCIAC0468-10ML TCIAC0468-25ML
1,4-Cyclohexadiene ≥98.0% stabilised
1,4-Cyclohexadiene
Formel: C₆H₈
MW: 80,13 g/mol
Kokpunkt: 88°C
Smältpunkt: -49°C
Densitet: 0,847 g/cm³ (20 °C)
Flampunkt: -11°C
Storage Temperature: Kylskåp
MDL Number: MFCD00001535
CAS nummer: 628-41-1
EINECS: 211-043-1
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ed860379852b0debcbf1d4dc93111abaf7a719d&vid=8ed860379852b0debcbf1d4dc93111abaf7a719d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR