2-Methyl-4-nitrophenol ≥98.0%

Leverantör: TCI
Varning

N0670-25G N0670-5G
TCIAN0670-25GEA 2575 SEK
TCIAN0670-25G TCIAN0670-5G
2-Methyl-4-nitrophenol ≥98.0%
2-Methyl-4-nitrophenol
Formel: C₇H₇NO₃
Kokpunkt: 311 °C (1013 hPa)
Smältpunkt: 96 °C
Storage Temperature: Omgivande
MDL Number: MFCD01734013
CAS nummer: 99-53-6
EINECS: 202-763-7
UN: 2446
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a704a0557f3ef29287910f6acc01cafb37c19fef&vid=221bacd649e22c45b50a0c504a6315215bd4aad9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Amber powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 93.0 to 97.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR