Disulfiram ≥97.0%
Leverantör: TCI
Varning
Synonyms: Tetraetyltiuramdisulfid
B0479-25G B0479-500G
TCIAB0479-25GEA 230 SEK
TCIAB0479-25G TCIAB0479-500G
Disulfiram ≥97.0%
Disulfiram
Formel: C₁₀H₂₀N₂S₄
Kokpunkt: 369 °C (1013 hPa)
Smältpunkt: 70 °C
Densitet: 1,34 g/cm³ (20 °C)
MDL Number: MFCD00009048
CAS nummer: 97-77-8
EINECS: 202-607-8
UN: 3077
ADR: 9,III
Merck Index: 14,03364
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8b58b436e004291aceefd763b0cd9a00f5b908ac&vid=8b58b436e004291aceefd763b0cd9a00f5b908ac&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (Iodometric Titration) min. 97.0 %
Melting point 69.0 to 73.0 deg-C