Isatin ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 2,3-Indolinedione, Indoline-2,3-dione, 1H-Indole-2,3-dione
I0080-25G I0080-500G I0080-100G
TCIAI0080-25GEA 295 SEK
TCIAI0080-25G TCIAI0080-500G TCIAI0080-100G
Isatin ≥98.0% (by GC, titration analysis)
Isatin
Formel: C₈H₅NO₂
MW: 147,13 g/mol
Kokpunkt: 342 °C (1013 hPa)
Smältpunkt: 198…202 °C
Densitet: 1,472 g/cm³ (20 °C)
Flampunkt: 220 °C
Storage Temperature: Omgivande
MDL Number: MFCD00005718
CAS nummer: 91-56-5
EINECS: 202-077-8
Merck Index: 14,05104
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4d5f20ce6e460f08a90897432ba5c63e36e31461&vid=4d5f20ce6e460f08a90897432ba5c63e36e31461&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 200.0 to 204.0 deg-C
Solubility in hot EtOH (95vol%) almost transparency