Solid phase extraction cartridges, Bond Elut™ PRS
Leverantör: VARIAN
12113038 12102012 12102039 12256010 12113012 12102074 12102094
VARI12113038EA 6125 SEK
VARI12113038 551-0308 VARI12102039 VARI12256010 VARI12113012 VARI12102074 VARI12102094
Solid phase extraction cartridges, Bond Elut™ PRS
Kromatografikolonn
Bond Elut™ PRS is a strong cation exchange sorbent that is also relatively high in polarity. With no appreciable degree of hydrophobicity, in non-polar solvents, PRS is capable of polar and hydrogen bonding interactions. Due to the very low pKa of PRS, a high ionic strength eluent must be used, and so the use of PRS is recommended for weaker cationic species such as pyridinium compounds.
  • Capable of polar and hydrogen bonding interactions
  • No appreciable non-polar interactions
  • Unique selectivity properties
Order Now
SPECIFIKATIONER
Environmentally Preferable
Kolbelastning 1,7%
Endcapped Nej
Surface area 500 m²/g