1-Cyclohexene-1-carboxylic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Cyclohex-1-enecarboxylic acid

C1296-1G C1296-5G
TCIAC1296-1GEA 415 SEK
TCIAC1296-1G TCIAC1296-5G
1-Cyclohexene-1-carboxylic acid ≥98.0%
1-Cyclohexene-1-carboxylic acid
Formel: C₇H₁₀O₂
MW: 126,16 g/mol
MDL Number: MFCD00001545
CAS nummer: 636-82-8
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f664596eb7de7f59eb065c1ef3e5e7739d3c5cef&vid=f664596eb7de7f59eb065c1ef3e5e7739d3c5cef&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorless to Light orange to Yellow powder to lump to clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR