Methyl cyclopropanecarboxylate 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Methylcyclopropanecarboxylate
126710250 126711000 126715000
ACRO126710250EA 635 SEK
ACRO126710250 ACRO126711000 ACRO126715000
Methyl cyclopropanecarboxylate 98%
Methyl cyclopropanecarboxylate
Formel: C₅H₈O₂
MW: 100,12 g/mol
Kokpunkt: 119 °C (764 mmHg)
Densitet: 0,98 g/cm³
Flampunkt: 17 °C
Storage Temperature: Omgivande
MDL Number: MFCD00001280
CAS nummer: 2868-37-3
EINECS: 220-690-9
UN: 1992
ADR: 3,II
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4170 to 1.4190 (20°C, 589 nm)
GC ≥97.5 %