Ethyl succinyl chloride ≥95.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Ethyl 4-chloro-4-oxobutanoate

E0368-25G E0368-500G
TCIAE0368-25GEA 590 SEK
TCIAE0368-25G TCIAE0368-500G
Ethyl succinyl chloride ≥95.0%
Ethyl succinyl chloride
Formel: C₆H₉ClO₃
MW: 164,59 g/mol
Kokpunkt: 144°C
Flampunkt: 84°C
MDL Number: MFCD00000751
CAS nummer: 14794-31-1
EINECS: 238-855-9
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f373ff8841676a6b75e874ad401e3f44957e0d6c&vid=f373ff8841676a6b75e874ad401e3f44957e0d6c&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (Argentmetric Titration) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR