Ethyl succinyl chloride 95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Ethyl 4-chloro-4-oxobutanoate
184090250 184091000
ACRO184090250EA 750 SEK
ACRO184090250 ACRO184091000
Ethyl succinyl chloride 95%
Ethyl succinyl chloride
Formel: C₆H₉ClO₃
Kokpunkt: 88…90 °C (11 mmHg)
Densitet: 1,1575 g/cm³
Flampunkt: 84 °C
Storage Temperature: Omgivande
MDL Number: MFCD00000751
CAS nummer: 14794-31-1
EINECS: 238-855-9
UN: 1760
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4350 to 1.4390 (20°C, 589 nm)
GC ≥94 %
Titration RCOCl ≥94 %