L-(+)-Methyl-tert-leucinate hydrochloride ≥98.0%

Leverantör: TCI EUROPE

Synonyms: L-tert-Leucine methyl ester hydrochloride, H-Tle-OMe·HCl, H-Tle-OMe.HCl

L0188-1G L0188-25G L0188-5G
TCIAL0188-1GEA 215 SEK
TCIAL0188-1G TCIAL0188-25G TCIAL0188-5G
L-(+)-Methyl-tert-leucinate hydrochloride ≥98.0%
L-(+)-Methyl-tert-leucinate hydrochloride
Formel: C₇H₁₅NO₂·HCl
MW: 181,7 g/mol
MDL Number: MFCD00040604
CAS nummer: 63038-27-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a4308cca0f1ca957d84ed3a033104ab96c0e716&vid=2a4308cca0f1ca957d84ed3a033104ab96c0e716&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +13.0 to +20.0 deg (C=1, Methanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR