4,6-Dimethyl-2-mercaptopyrimidine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Mercapto-4,6-dimethylpyrimidine
D1580-25G
D1580-250G
TCIAD1580-25GEA
565
SEK
TCIAD1580-25G
TCIAD1580-250G
4,6-Dimethyl-2-mercaptopyrimidine ≥98.0%
4,6-Dimethyl-2-mercaptopyrimidine
Formel:
C₆H₈N₂S MW: 140,20 g/mol |
MDL Number:
MFCD00006079 CAS nummer: 22325-27-5 EINECS: 244-911-3 |
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...