Dibenzothiophene-4-boronic acid (contains varying amounts of Anhydride)
Leverantör: TCI
|
Varning
|
D4057-1G
D4057-5G
TCIAD4057-1GEA
215
SEK
TCIAD4057-1G
TCIAD4057-5G
Dibenzothiophene-4-boronic acid (contains varying amounts of Anhydride)
Dibenzothiophene-4-boronic acid
Formel:
C₁₂H₉BO₂S Smältpunkt: 339°C |
MDL Number:
MFCD01318182 CAS nummer: 108847-20-7 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 97.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...