Benzofuran-2-carboxylic acid ≥99.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Benzo[b]furan-2-carboxylic acid
B1981-1G
B1981-5G
TCIAB1981-1GEA
310
SEK
TCIAB1981-1G
TCIAB1981-5G
Benzofuran-2-carboxylic acid ≥99.0% (by GC, titration analysis)
Benzofuran-2-carboxylic acid
Formel:
C₉H₆O₃ MW: 162,15 g/mol |
MDL Number:
MFCD00005848 CAS nummer: 496-41-3 EINECS: 207-818-9 Merck Index: 14,02561 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 193.0 to 197.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...