4-Chloro-DL-phenylalanine 98%
Leverantör: Thermo Scientific
Synonyms: p-Chloro-DL-Phenylalanine, H-DL-Phe(4-Cl)-OH
157280050 157280250
ACRO157280050EA 1200 SEK
ACRO157280050 ACRO157280250
4-Chloro-DL-phenylalanine 98%
4-Chloro-DL-phenylalanine
Formel: C₉H₁₀ClNO₂
MW: 199,6 g/mol
Smältpunkt: >240 °C
Storage Temperature: Omgivande
MDL Number: MFCD00002601
CAS nummer: 7424-00-2
EINECS: 231-051-9
Order Now
Specifikation testresultat
Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
HPLC ≥97.5 %
Titration with HClO4 ≥97.5 %