1,3-Propylenediphosphonic acid ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Propylenediphosphonic acid
30277.02 30277.04 30277.09
30277.02EA 1600 SEK
30277.02 30277.04 30277.09
1,3-Propylenediphosphonic acid ≥98%
1,3-Propylenediphosphonic acid
Purity: 98+% CAS No.: 4671-82-3 Molecular Formula: [P(O)(OH)2CH2]2CH2 Formula Weight: 204.06 Physical Form: Powder Odour: Odourless MDL No.: MFCD00050066
Formel: C₃H₁₀O₆P₂
MW: 204,06 g/mol
Smältpunkt: 178 °C
Storage Temperature: Omgivande
MDL Number: MFCD00050066
CAS nummer: 4671-82-3