1-(2-Aminoethyl)pyrrolidine 99%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: N-(2-Aminoethyl)pyrrolidine, Pyrrolidinoethylamine
103700050 103700250 103701000
ACRO103700050EA 820 SEK
ACRO103700050 ACRO103700250 ACRO103701000
1-(2-Aminoethyl)pyrrolidine 99%
1-(2-Aminoethyl)pyrrolidine
Formel: C₆H₁₄N₂
Kokpunkt: 66…70 °C (23 mmHg)
Densitet: 0,9 g/cm³
Flampunkt: 47 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003182
CAS nummer: 7154-73-6
EINECS: 230-509-5
UN: 1993
ADR: 3,III
Order Now
Specifikation testresultat
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4677 to 1.4697 (20°C, 589 nm)
GC ≥88 %