trans-4-Butylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

B1136-25G B1136-5G
TCIAB1136-25GEA 1700 SEK
TCIAB1136-25G TCIAB1136-5G
trans-4-Butylcyclohexanecarboxylic acid ≥99.0% (by GC, titration analysis)
trans-4-Butylcyclohexanecarboxylic acid
Formel: C₁₁H₂₀O₂
MW: 184,28 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00059560
CAS nummer: 38289-28-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b33138a6c5ec1bef8929fac84f4a142781bbace5&vid=b33138a6c5ec1bef8929fac84f4a142781bbace5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Purity (Neutralization titration) min. 99.0 %
Solubility in Methanol almost transparency
Melting point 35.0 to 38.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR