5-Benzoylpentanoic acid ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
6-Oxo-6-phenylhexanoic acid
B2182-25G
B2182-5G
TCIAB2182-25GEA
2750
SEK
TCIAB2182-25G
TCIAB2182-5G
5-Benzoylpentanoic acid ≥98.0% (by GC, titration analysis)
5-Benzoylpentanoic acid
Formel:
C₁₂H₁₄O₃ MW: 206,24 g/mol |
MDL Number:
MFCD00014380 CAS nummer: 4144-62-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 76.0 to 79.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...