tert-Butyl-4-aminobenzoate ≥98.0% (by GC, titration analysis)
Leverantör: TCI
Varning
Synonyms: tert-Butyl 4-aminobenzoate
B3504-25G B3504-5G
TCIAB3504-25GEA 1875 SEK
TCIAB3504-25G TCIAB3504-5G
tert-Butyl-4-aminobenzoate ≥98.0% (by GC, titration analysis)
tert-Butyl-4-aminobenzoate
Formel: C₁₁H₁₅NO₂
MW: 193.2423 g/mol
MDL Number: MFCD00665790
CAS nummer: 18144-47-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca0c1e64d03530ec45d13b144a26db011e5002b8&vid=ca0c1e64d03530ec45d13b144a26db011e5002b8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 108.0 to 112.0 deg-C