2-Cyanopyrazine ≥99.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Pyrazine-2-carbonitrile, 2-Pyrazinecarbonitril, Pyrazinecarbonitrile
C1123-25G
C1123-500G
TCIAC1123-25GEA
230
SEK
TCIAC1123-25G
TCIAC1123-500G
2-Cyanopyrazine ≥99.0%
2-Cyanopyrazine
Formel:
C₅H₃N₃ MW: 105,10 g/mol Kokpunkt: 87°C |
MDL Number:
MFCD00049361 CAS nummer: 19847-12-2 EINECS: 243-369-5 ADR: 6.1,III |
Specifikation testresultat
Appearance | White or Colorless to Light yellow powder to lump to clear liquid |
Purity (GC) | min. 99.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...