Diethylbenzene (mixture of isomers) ≥98.0%

Leverantör: TCI
Fara

D3378-25ML D3378-500ML
TCIAD3378-25MLEA 215 SEK
TCIAD3378-25ML TCIAD3378-500ML
Diethylbenzene (mixture of isomers) ≥98.0%
Diethylbenzene (mixture of isomers)
Formel: C₁₀H₁₄
Kokpunkt: 184°C
Smältpunkt: –31°C
Flampunkt: 55°C
MDL Number: MFCD00792900
CAS nummer: 25340-17-4
EINECS: 246-874-9
UN: 2049
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomers)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR