4-Nitrophthalhydrazide ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI
Synonyms:
4-Nitrophthalhydrazide, 2,3-Dihydro-6-nitro-1,4-phthalazinedione, 4-Nitrophthalic Hydrazide
N0799-25G
N0799-5G
TCIAN0799-25GEA
1475
SEK
TCIAN0799-25G
TCIAN0799-5G
4-Nitrophthalhydrazide ≥98.0% (by HPLC, titration analysis)
4-Nitrophthalhydrazide
Formel:
C₈H₅N₃O₄ Smältpunkt: 300°C |
MDL Number:
MFCD00006891 CAS nummer: 3682-19-7 |
|
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...