Phthalan ≥97.0%
Leverantör: TCI
Synonyms:
1,3-Dihydrobenzo[c]furan
X0053-25G
X0053-5G
TCIAX0053-25GEA
6800
SEK
TCIAX0053-25G
TCIAX0053-5G
Phthalan ≥97.0%
Phthalan
Formel:
C₈H₈O Flampunkt: 63°C |
MDL Number:
MFCD00005932 CAS nummer: 496-14-0 EINECS: 207-815-2 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...