Perfluorobutane-1-sulphonic acid ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: Perfluoro-1-butanesulphonic acid, Nonafluoro-1-butanesulphonic acid

N0709-25G N0709-5G
TCIAN0709-25GEA 1975 SEK
TCIAN0709-25G TCIAN0709-5G
Perfluorobutane-1-sulphonic acid ≥98.0%
Perfluorobutane-1-sulphonic acid
Formel: C₄HF₉O₃S
Kokpunkt: 212°C
MDL Number: MFCD01320794
CAS nummer: 375-73-5
EINECS: 206-793-1
UN: 2586
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90145d3b2283f6a8732960070ad272dbec5fa3c1&vid=90145d3b2283f6a8732960070ad272dbec5fa3c1&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR