Norbornadiene ≥97.0% stabilised
Leverantör: TCI
|
Fara
|
Synonyms:
2,5-Norbornadiene
Stabilization:
Stabilised med BHT/Ionol
N0346-25ML
N0346-500ML
N0346-100ML
TCIAN0346-25MLEA
670
SEK
TCIAN0346-25ML
TCIAN0346-500ML
TCIAN0346-100ML
Norbornadiene ≥97.0% stabilised
Norbornadiene
Formel:
C₇H₈ MW: 92,14 g/mol Kokpunkt: 90°C Flampunkt: –19°C |
MDL Number:
MFCD00082301 CAS nummer: 121-46-0 EINECS: 204-472-0 UN: 2251 ADR: 3,II |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...