5-(4-(Diethylamino)benzylidene)rhodanine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
(5E)-5-[4-(Diethylamino)benzylidene]-2-mercapto-1,3-thiazol-4(5H)-one
D0464-1G
D0464-5G
TCIAD0464-1GEA
215
SEK
TCIAD0464-1G
TCIAD0464-5G
5-(4-(Diethylamino)benzylidene)rhodanine ≥98.0%
5-(4-(Diethylamino)benzylidene)rhodanine
Formel:
C₁₄H₁₆N₂OS₂ |
MDL Number:
MFCD00193758 CAS nummer: 35778-58-6 EINECS: 252-723-8 |
Specifikation testresultat
Appearance | Red to Dark red to Dark purple powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in hot Acetic acid | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...