2-Vinylpyridine ≥97.0% stabilised

Leverantör: TCI
Fara


Stabilization: med 4-tert-Butylpyrokatekol

V0024-25ML V0024-500ML
TCIAV0024-25MLEA 215 SEK
TCIAV0024-25ML TCIAV0024-500ML
2-Vinylpyridine ≥97.0% stabilised
2-Vinylpyridine
Formel: C₇H₇N
Kokpunkt: 158°C
Smältpunkt: –50 °C
Densitet: 0,9735 g/cm³ (20 °C)
Flampunkt: 46°C
Storage Temperature: Frys
MDL Number: MFCD00006355
CAS nummer: 100-69-6
EINECS: 202-879-8
UN: 3073
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1d6c5a94391def261b11e9fbe1b1c8bdfd80f35&vid=e1d6c5a94391def261b11e9fbe1b1c8bdfd80f35&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Brown clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR