1,4-Butandioldiakrylat ≥90.0% stabilised

Leverantör: TCI EUROPE
Fara


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

B2935-25G B2935-500G B2935-100G
TCIAB2935-25GEA 270 SEK
TCIAB2935-25G TCIAB2935-500G TCIAB2935-100G
1,4-Butandioldiakrylat ≥90.0% stabilised
1,4-Butandioldiakrylat
Formel: C₁₀H₁₄O₄
MW: 198,22 g/mol
Kokpunkt: 138°C
MDL Number: MFCD00014940
CAS nummer: 1070-70-8
EINECS: 213-979-6
UN: 1760
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2d00eaf166daf35d4d8dd2c60c746df70aaedc8&vid=a2d00eaf166daf35d4d8dd2c60c746df70aaedc8&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR