Tetramethyl-1,3-cyclobutanedione ≥99.0% precursor to dimethyl ketene

Leverantör: TCI EUROPE

Synonyms: Tetramethylcyclobutane-1,3-dione

D0258-25G D0258-5G
TCIAD0258-25GEA 1600 SEK
TCIAD0258-25G TCIAD0258-5G
Tetramethyl-1,3-cyclobutanedione ≥99.0% precursor to dimethyl ketene
Tetramethyl-1,3-cyclobutanedione
Formel: C₈H₁₂O₂
MW: 140,18 g/mol
MDL Number: MFCD00001331
CAS nummer: 933-52-8
EINECS: 213-269-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4b773139062f343980eb2e50930e6af9d7d4297&vid=9d21592fb9c680852f825e13671c6910e3d5b027&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 112.0 to 115.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR