1-(2-(Trifluoromethyl)phenyl)piperazine ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

T2739-1G T2739-5G
TCIAT2739-1GEA 1400 SEK
TCIAT2739-1G TCIAT2739-5G
1-(2-(Trifluoromethyl)phenyl)piperazine ≥98.0% (by GC, titration analysis)
1-(2-(Trifluoromethyl)phenyl)piperazine
Formel: C₁₁H₁₃F₃N₂
MDL Number: MFCD02094727
CAS nummer: 63854-31-9
EINECS: 264-514-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b944c35efda0f0e286b7ac6034eb89a611b83bed&vid=b944c35efda0f0e286b7ac6034eb89a611b83bed&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR