Vinyl laurate ≥99.0% stabilised

Leverantör: TCI

Synonyms: Lauric acid Vinyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

L0090-25ML L0090-500ML
TCIAL0090-25MLEA 215 SEK
TCIAL0090-25ML TCIAL0090-500ML
Vinyl laurate ≥99.0% stabilised
Vinyl laurate
Formel: C₁₄H₂₆O₂
Flampunkt: 136°C
MDL Number: MFCD00048421
CAS nummer: 2146-71-6
EINECS: 218-414-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=02e39cdef48c1aea6f60cfd54a5f4b51fc438e3a&vid=02e39cdef48c1aea6f60cfd54a5f4b51fc438e3a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR