3-Quinolinecarbaldehyde ≥96.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: Quinoline-3-carboxaldehyde, Quinoline-3-carbaldehyde, 3-Quinolinecarboxaldehyde

Q0075-1G Q0075-5G
TCIAQ0075-1GEA 845 SEK
TCIAQ0075-1G TCIAQ0075-5G
3-Quinolinecarbaldehyde ≥96.0% (by GC, titration analysis)
3-Quinolinecarbaldehyde
Formel: C₁₀H₇NO
MW: 157,17 g/mol
Kokpunkt: 128°C
Storage Temperature: Kylskåp
MDL Number: MFCD00006768
CAS nummer: 13669-42-6
EINECS: 237-147-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ebf08128e2d3164d850a1d415cf8bbedf5fe591e&vid=7724329c1387547c596cba69fdc80b2ddb03be39&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 96.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 66.0 to 71.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR