N-Benzylaniline ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
N-Phenylbenzylamine, ((Phenylmethyl)amino)benzene
P0159-25G
P0159-500G
P0159-100G
TCIAP0159-25GEA
310
SEK
TCIAP0159-25G
TCIAP0159-500G
TCIAP0159-100G
N-Benzylaniline ≥98.0%
N-Benzylaniline
Formel:
C₆H₅CH₂NHC₆H₅ MW: 183,25 g/mol Kokpunkt: 307°C Smältpunkt: 33…36 °C Densitet: 1,06 g/cm³ (20 °C) Flampunkt: 152 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00003018 CAS nummer: 103-32-2 EINECS: 203-100-4 Merck Index: 14,01126 |
Specifikation testresultat
Appearance | White or Colorless to Yellow powder to lump to clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Solubility in Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...