trans-2-Fluorocyclohexanol ≥98.0%
Leverantör: TCI
F0631-1G
F0631-5G
TCIAF0631-1GEA
1100
SEK
TCIAF0631-1G
TCIAF0631-5G
trans-2-Fluorocyclohexanol ≥98.0%
trans-2-Fluorocyclohexanol
Formel:
C₆H₁₁FO Kokpunkt: 166°C Smältpunkt: 21°C Storage Temperature: Kylskåp |
MDL Number:
MFCD08146648 CAS nummer: 656-60-0 |
|
Specifikation testresultat
Appearance | White to Almost white powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...