N, N-dimetylanilin ≥99.0% for biochemical research
Leverantör: TCI
Fara
D3866-1G D3866-5G
TCIAD3866-1GEA 215 SEK
TCIAD3866-1G TCIAD3866-5G
N, N-dimetylanilin ≥99.0% for biochemical research
N, N-dimetylanilin
Formel: C₈H₁₁N
MW: 121.18 g/mol
Kokpunkt: 194°C
Smältpunkt: 2,5
Flampunkt: 73°C
Storage Temperature: Omgivande
MDL Number: MFCD00008304
CAS nummer: 121-69-7
EINECS: 204-493-5
UN: 2253
ADR: 6.1 II
Merck Index: 14,03234
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97ecaacb6f7a3f03a7c1ed26bca2830feeecf7fd&vid=7cdc43992ac3f330e64cc9a963c9d3c3dbca1581&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 99.0 %
Refractive index n20/D 1.5560 to 1.5600
Hydrogendioxide detection test (Peroxidase method) to pass test